siRNA Details
Gene Symbol: LOC440602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108042.2 | 2 | 590 | |||
GenBank mRNA | BC042100.1 | - | 2 | 590 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ027751.1 | - | 3 | 484 | Chr. 1 - 110489357 - 110491272 [-] on Build GRCh38 |
