siRNA Details
Gene Symbol: LOC100130744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129556.1 | - | 1 | 764 |
Gene Symbol: OTULIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514151.2 | XP_011512453.1 | 8 | 7859 |
