siRNA Details
Gene Symbol: ZNF433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308351.1 | NP_001295280.1 | 2 | 221 | ||
NM_001308357.1 | NP_001295286.1 | 2 | 221 | |||
XM_011527728.2 | XP_011526030.1 | 2 | 102 | |||
XM_011527729.2 | XP_011526031.1 | 2 | 102 | |||
GenBank mRNA | BC047412.1 | - | 2 | 156 | 5295132 | |
BI831553.1 | - | 2 | 176 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129553.1 | - | 1 | 818 | Chr. 19 - 12032836 - 12035676 [-] on Build GRCh38 |
