siRNA Details
Gene Symbol: CLEC19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256720.1 | NP_001243649.1 | 5 | 768 | ||
GenBank mRNA | BX640722.1 | - | 3 | 655 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CLEC19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256720.1 | NP_001243649.1 | 5 | 768 | ||
GenBank mRNA | BX640722.1 | - | 3 | 655 |