siRNA Details
Gene Symbol: MIATNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026502.1 | - | 2 | 1173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN361213.1 | - | 3 | 505 | Chr. 22 - 26672856 - 26718392 [+] on Build GRCh38 |
