siRNA Details
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AJ412050.1 | - | LIT1890 BCMS |
2 | 189 | |
AJ412051.1 | - | LIT1889 BCMS |
1 | 110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AJ411990.3 | - | 1 | 110 | Chr. 13 - 50389087 - 50389552 [+] on Build GRCh38 |
