siRNA Details
Gene Symbol: LOC100128651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055457.1 | - | 1 | 1672 | ||
AK127243.1 | - | 1 | 3439 | |||
D25271.1 | - | 1 | 1160 |
Gene Symbol: C4orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513603.2 | XP_011511905.1 | 11 | 13699 | ||
XM_011513604.2 | XP_011511906.1 | 12 | 13965 | |||
XM_017008891.1 | XP_016864380.1 | 14 | 13631 | |||
XM_017008892.1 | XP_016864381.1 | 12 | 14339 | |||
XM_017008893.1 | XP_016864382.1 | 13 | 13464 |
