siRNA Details
Gene Symbol: LOC100128651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055457.1 | - | 1 | 281 | ||
AK127243.1 | - | 1 | 2048 |
Gene Symbol: C4orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513603.2 | XP_011511905.1 | 11 | 12308 | ||
XM_011513604.2 | XP_011511906.1 | 12 | 12574 | |||
XM_017008891.1 | XP_016864380.1 | 14 | 12240 | |||
XM_017008892.1 | XP_016864381.1 | 12 | 12948 | |||
XM_017008893.1 | XP_016864382.1 | 13 | 12073 |
