siRNA Details
Gene Symbol: LOC100128651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127243.1 | - | 1 | 1635 |
Gene Symbol: C4orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513603.2 | XP_011511905.1 | 11 | 11895 | ||
XM_011513604.2 | XP_011511906.1 | 12 | 12161 | |||
XM_017008891.1 | XP_016864380.1 | 14 | 11827 | |||
XM_017008892.1 | XP_016864381.1 | 12 | 12535 | |||
XM_017008893.1 | XP_016864382.1 | 13 | 11660 |
