siRNA Details
Gene Symbol: SMARCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003601.3 | NP_003592.3 | 24 | 4148 | ||
GenBank mRNA | AL157495.1 | - | 1 | 266 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM473431.1 | - | 1 | 77 | Chr. 4 - 143553639 - 143554364 [+] on Build GRCh38 |
