siRNA Details
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094351.1 | - | 1 | 639 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD523126.1 | - | 1 | 37 | Chr. 16 - 9105452 - 9106160 [+] on Build GRCh38 |
