siRNA Details
Gene Symbol: LOC105372321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627135.1 | - | 1 | 1691 |
Gene Symbol: LOC220729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309924.1 | - | 7 | 924 |
Gene Symbol: LOC283045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104162.1 | 4 | 670 | |||
GenBank mRNA | AK095019.1 | - | 4 | 671 |
Gene Symbol: LOC400706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125344.1 | 2 | 722 | |||
GenBank mRNA | AK096622.1 | - | 2 | 722 |
