siRNA Details
Gene Symbol: ZNF677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA592734.1 | - | 4 | 499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AX721159.1 | - | 1 | 402 | Chr. 19 - 53242211 - 53244011 [-] on Build GRCh38 |
