siRNA Details
Gene Symbol: LOC101927683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110683.1 | 4 | 1824 | |||
GenBank mRNA | AK093200.1 | - | 4 | 1824 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110683.1 | 4 | 1824 | |||
GenBank mRNA | AK093200.1 | - | 4 | 1824 |