siRNA Details
Gene Symbol: COLCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017197.1 | XP_016872686.1 | 1 | 1011 | ||
GenBank mRNA | AB096246.1 | - | 1 | 1456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM821976.1 | - | 1 | 109 | Chr. 11 - 111303379 - 111303887 [+] on Build GRCh38 |
