siRNA Details
Gene Symbol: TMEM181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536003.2 | XP_011534305.1 | 11 | 4790 | ||
GenBank mRNA | AK023059.1 | - | 1 | 1761 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536003.2 | XP_011534305.1 | 11 | 4790 | ||
GenBank mRNA | AK023059.1 | - | 1 | 1761 |