siRNA Details
Gene Symbol: LINC00572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047501.1 | 2 | 241 | |||
GenBank mRNA | BC039674.1 | - | 2 | 240 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047501.1 | 2 | 241 | |||
GenBank mRNA | BC039674.1 | - | 2 | 240 |