siRNA Details
Gene Symbol: LOC102723448 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110824.1 | 2 | 75 | |||
GenBank mRNA | BC039672.1 | - | 2 | 75 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI826035.1 | - | 2 | 74 | Chr. 3 - 11749 - 24498 [+] on Build GRCh38 |
