siRNA Details
Gene Symbol: LINC00558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047488.1 | 6 | 3016 | |||
GenBank mRNA | BC041877.1 | - | 6 | 3016 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047488.1 | 6 | 3016 | |||
GenBank mRNA | BC041877.1 | - | 6 | 3016 |