siRNA Details
Gene Symbol: LOC101928823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120559.1 | 3 | 761 | |||
GenBank mRNA | BC029570.1 | - | 3 | 657 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI545849.1 | - | 3 | 661 | Chr. 11 - 112393116 - 112541911 [+] on Build GRCh38 |
