siRNA Details
Gene Symbol: BOLA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045634.1 | 2 | 3651 | |||
NR_045636.1 | 3 | 3525 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK123865.1 | - | 1 | 613 | Chr. 2 - 74151238 - 74154192 [+] on Build GRCh38 |
