siRNA Details
Gene Symbol: LOC100505736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120504.1 | 1 | 292 | |||
GenBank mRNA | AK057415.1 | - | 1 | 295 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB022519.1 | - | 1 | 292 | Chr. 2 - 28151188 - 28206961 [-] on Build GRCh38 |
