siRNA Details
Gene Symbol: LOC285804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126021.1 | 3 | 382 | |||
GenBank mRNA | AK097676.1 | - | 3 | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB045863.1 | - | 3 | 382 | Chr. 6 - 170268063 - 170276762 [-] on Build GRCh38 |
