siRNA Details
Gene Symbol: LOC284926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091355.1 | - | 2 | 1274 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR624447.1 | - | 1 | 1469 | Chr. 22 - 46250437 - 46253237 [+] on Build GRCh38 |
