siRNA Details
Gene Symbol: LOC101928107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109961.1 | 2 | 264 | |||
GenBank mRNA | BC021745.1 | - | 3 | 269 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG718233.1 | - | 3 | 269 | Chr. 21 - 33112321 - 33123700 [-] on Build GRCh38 |
