siRNA Details
Gene Symbol: LOC101928107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109961.1 | 3 | 1080 | |||
GenBank mRNA | BC021745.1 | - | 4 | 1085 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109961.1 | 3 | 1080 | |||
GenBank mRNA | BC021745.1 | - | 4 | 1085 |