siRNA Details
Gene Symbol: C1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297721.1 | NP_001284650.1 | 5 | 1517 | ||
NM_001297723.1 | NP_001284652.1 | 6 | 1619 | |||
GenBank mRNA | AK290583.1 | - | 6 | 1391 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC017002.1 | - | 1041 | Chr. - - [-] on Build |
