siRNA Details
Gene Symbol: LOC101929454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109944.1 | 3 | 510 | |||
GenBank mRNA | BC002741.2 | - | 3 | 510 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE394211.1 | - | 3 | 492 | Chr. 5 - 15192139 - 15266541 [-] on Build GRCh38 |
