siRNA Details
Gene Symbol: LOC101928371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110236.1 | 1 | 23 | |||
GenBank mRNA | BC044304.1 | - | 1 | 23 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ423198.1 | - | 1 | 21 | Chr. 2 - 88538722 - 88574644 [+] on Build GRCh38 |
