siRNA Details
Gene Symbol: MIR124-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109792.1 | 3 | 2261 | |||
GenBank mRNA | AK124256.1 | - | 3 | 2262 | ||
BX537900.1 | - | 2 | 2155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX537518.1 | - | 1 | 3974 | Chr. 8 - 64379233 - 64383923 [+] on Build GRCh38 |
