siRNA Details
Gene Symbol: SIGMAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC006307.1 | - | 1153 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE261484.1 | - | 1 | 181 | Chr. 22 - 41674946 - 41675561 [-] on Build GRCh38 |
