siRNA Details
Gene Symbol: LOC646522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120444.1 | 3 | 557 | |||
GenBank mRNA | BC043577.2 | - | 2 | 530 | ||
BI836817.1 | - | 3 | 557 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC646522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120444.1 | 3 | 557 | |||
GenBank mRNA | BC043577.2 | - | 2 | 530 | ||
BI836817.1 | - | 3 | 557 |