siRNA Details
Gene Symbol: ZNF213-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF098968.1 | - | 2 | 554 | ||
BC013599.1 | - | 1 | 217 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF213-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF098968.1 | - | 2 | 554 | ||
BC013599.1 | - | 1 | 217 |