siRNA Details
Gene Symbol: LOC101928135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110817.1 | 3 | 235 | |||
GenBank mRNA | BC035904.1 | - | 3 | 235 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI918059.1 | - | 3 | 235 | Chr. 3 - 34875992 - 35394023 [-] on Build GRCh38 |
