siRNA Details
Gene Symbol: LZTS1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047509.1 | 1 | 42 | |||
GenBank mRNA | BC038538.1 | - | 1 | 42 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI830862.1 | - | 1 | 42 | Chr. 8 - 20275773 - 20290431 [+] on Build GRCh38 |
