siRNA Details
Gene Symbol: LOC101928140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125849.1 | 2 | 107 | |||
GenBank mRNA | BC033773.1 | - | 2 | 118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE894759.1 | - | 86 | Chr. - - [-] on Build |
