siRNA Details
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046357.1 | 2 | 1345 | |||
GenBank mRNA | AK055997.1 | - | 3 | 1479 | ||
AK074561.1 | - | 2 | 1370 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046357.1 | 2 | 1345 | |||
GenBank mRNA | AK055997.1 | - | 3 | 1479 | ||
AK074561.1 | - | 2 | 1370 |