siRNA Details
Gene Symbol: KCNK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104210.1 | 4 | 2683 | |||
XM_011517101.2 | XP_011515403.1 | 4 | 2438 | |||
XM_011517102.2 | XP_011515404.1 | 4 | 4634 | |||
XM_011517103.2 | XP_011515405.1 | 4 | 2182 | |||
XM_017013530.1 | XP_016869019.1 | 4 | 4350 | |||
XM_017013531.1 | XP_016869020.1 | 4 | 2449 | |||
GenBank mRNA | AK090707.1 | - | 3 | 2116 | ||
AK094401.1 | - | 1 | 1679 | |||
BC034734.1 | - | 3 | 1529 |
