siRNA Details
Gene Symbol: LOC100505635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091770.1 | - | 1 | 730 | ||
AK125892.1 | - | 1 | 3093 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100505635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091770.1 | - | 1 | 730 | ||
AK125892.1 | - | 1 | 3093 |