siRNA Details
Gene Symbol: LOC100505635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125831.1 | 4 | 558 | |||
GenBank mRNA | AK125892.1 | - | 1 | 35 | ||
AL079669.1 | - | 4 | 533 | |||
DA315022.1 | - | 2 | 215 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100505635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125831.1 | 4 | 558 | |||
GenBank mRNA | AK125892.1 | - | 1 | 35 | ||
AL079669.1 | - | 4 | 533 | |||
DA315022.1 | - | 2 | 215 |