siRNA Details
Gene Symbol: LINC00619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033923.1 | 3 | 239 | |||
GenBank mRNA | BC017939.1 | - | 3 | 239 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF670044.1 | - | 3 | 239 | Chr. 10 - 43845306 - 43850199 [+] on Build GRCh38 |
