siRNA Details
Gene Symbol: TOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU677026.1 | - | 1 | 462 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124183.1 | - | 1 | 1507 | Chr. 14 - 21498139 - 21500234 [-] on Build GRCh38 |
