siRNA Details
Gene Symbol: LOC101928403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017749.1 | - | 1 | 2963 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF575623.1 | - | 1 | 383 | Chr. 2 - 88630750 - 88631455 [+] on Build GRCh38 |
