siRNA Details
Gene Symbol: LINC01115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033880.3 | 4 | 1400 | |||
NR_111963.1 | 3 | 1163 | ||||
GenBank mRNA | BC043553.1 | - | 4 | 1401 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033880.3 | 4 | 1400 | |||
NR_111963.1 | 3 | 1163 | ||||
GenBank mRNA | BC043553.1 | - | 4 | 1401 |