siRNA Details
Gene Symbol: LINC01115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033880.3 | 3 | 592 | |||
NR_111963.1 | 2 | 355 | ||||
GenBank mRNA | BC043553.1 | - | 3 | 593 | ||
BC128382.1 | - | 2 | 120 | |||
BC128532.1 | - | 2 | 120 |
