siRNA Details
Gene Symbol: ECHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541719.2 | XP_011540021.1 | 2 | 510 | ||
XM_017001638.1 | XP_016857127.1 | 3 | 899 | |||
XM_017001641.1 | XP_016857130.1 | 3 | 899 | |||
GenBank mRNA | AK309535.1 | - | 3 | 734 | ||
BX648522.1 | - | 1 | 1209 |
