siRNA Details
Gene Symbol: LOC101926984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040584.1 | - | 1 | 1070 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD557292.1 | - | 1 | 488 | Chr. 17 - 28265014 - 28265787 [-] on Build GRCh38 |
