siRNA Details
Gene Symbol: CCDC134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261748.3 | XP_005261805.1 | 8 | 4562 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124515.1 | - | 1 | 1873 | Chr. 22 - 41827573 - 41832162 [+] on Build GRCh38 |
