siRNA Details
Gene Symbol: LOC101929460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134622.1 | 2 | 379 | |||
GenBank mRNA | BC040917.1 | - | 2 | 379 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM919882.1 | - | 2 | 378 | Chr. 6 - 169034249 - 169035642 [-] on Build GRCh38 |
