siRNA Details
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306215.1 | NP_001293144.1 | 15 | 6949 | ||
NM_178835.4 | NP_849157.2 | 15 | 6958 | |||
XM_011531634.2 | XP_011529936.1 | 14 | 6963 | |||
XM_011531635.2 | XP_011529937.1 | 15 | 12450 | |||
XM_011531645.2 | XP_011529947.1 | 10 | 4701 | |||
XM_017007768.1 | XP_016863257.1 | 14 | 6958 | |||
XM_017007769.1 | XP_016863258.1 | 14 | 13209 | |||
XM_017007770.1 | XP_016863259.1 | 13 | 6730 | |||
XM_017007777.1 | XP_016863266.1 | 11 | 4700 | |||
GenBank mRNA | BC033248.1 | - | 1 | 2112 |
